Anti-Bcl-2 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Bcl-2 (phospho Ser70) for WB and ICC/IF with samples derived from Human and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Bcl-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Specifications
- Catalog No:
- 76850-040
- Antigen symbol:
- Bcl-2
- Antigen name:
- Apoptosis regulator Bcl 2
- Antigen synonyms:
- C430015F12Rik|AW986256|Apoptosis regulator Bcl2|D830018M01Rik|BCL2_HUMAN|Apoptosis regulator Bcl-2|B cell CLL/lymphoma 2|D630044D05Rik|B cell leukemia/lymphoma 2
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat
- Host:
- Rabbit
- Gene ID:
- UniprotID# P10415
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- Modification:
- phospho Ser70
- Immunogen:
- A synthetic phosphorylated peptide around S70 of human Bcl-2 (NP_000624.2)
- Sequence:
- RTSPL
- Form:
- Liquid
- Molecular weight:
- 28 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Tested applications:
- ICC/IF
Specifications
Frequently Bought Together