Anti-Guanylate kinase Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Guanylate kinase for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:200 to 1:1000
Type: Primary
Antigen: Guanylate kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 76851-742
- Antigen symbol:
- Guanylate kinase
- Antigen name:
- ATP:GMP phosphotransferase
- Antigen synonyms:
- GMP kinase|GUK1|Guanylate kinase|FLJ42686|GMK|FLJ43710|KGUA_HUMAN
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q16774
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 197 of human GUK1 (NP_000849.1)
- Sequence:
- MSGPRPVVLSGPSGAGKSTLLKRLLQEHSGIFGFSVSHTTRNPRPGEENGKDYYFVTREVMQRDIAAGDFIEHAEFSGNLYGTSKVAVQAVQAMNRICVLDVDLQGVRNIKATDLRPIYISVQPPSLHVLEQRLRQRNTETEESLVKRLAAAQADMESSKEPGLFDVVIINDSLDQAYAELKEALSEEIKKAQRTGA
- Form:
- Liquid
- Molecular weight:
- 21 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together