Anti-Histone H2B Rabbit Polyclonal Antibody
Catalog # 77063-224
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:H2B histone family
- Antigen symbol:Histone H2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:S. pombe
- Western blot:Yes
- Size:50 µl
- ChIP:Yes
- Form:Liquid
- Gene ID:UniprotID# P04913
- Antigen synonyms:member O|H2B 1A|H2B-GL105|H2B GL105|member Q|GL105|member S|H2B|H2B type 12
- Storage buffer:Supplied as antiserum with 0.05% Sodium Azide
- Sequence:SAAEKKPASKAPAGKA
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the N-terminal of Schizosaccharomyces pombe Histone H2B protein
- Cat. no.:77063-224
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Histone H2B for WB, IP and ChIP with samples derived from Schizosaccharomyces pombe.
- Validated Applications: WB, IP, ChIP
- Recommended Dilutions: WB: 1:1000
Type: Primary
Antigen: Histone H2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Schizosaccharomyces pombe