Anti-MAPRE3 Mouse Monoclonal Antibody [clone: KT113]
Catalog # 77041-658
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:APC binding protein
- Antigen symbol:MAPRE3
- Clonality:Monoclonal
- Clone:KT113
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UPY8
- Antigen synonyms:MAPRE 3|EBF3 S|End-binding protein 3|EBF3|EBF 3|End binding protein 3|EB1 protein family member 3|EB3|EB 3
- Storage buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:S162 phosphorylated EB3 peptide conjugated to carrier protein
- Purification:Antibody is purified from culture supernatant using a protein A column
- Cat. no.:77041-658
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal (KT113) antibody to MAPRE3 (phospho Ser162) for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1/500 to 1/20,000
Type: Primary
Antigen: MAPRE3
Clonality: Monoclonal
Clone: KT113
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human