Anti-GATA3 Rat Monoclonal Antibody [clone: KT63]
77041-580
- Antibody type:Primary
- Antigen name:GATA 3
- Antigen symbol:GATA3
- Clonality:Monoclonal
- Clone:KT63
- Conjugation:Unconjugated
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P23772
- Antigen synonyms:MGC5199|GATA3_HUMAN|HDRS|MGC2346|GATA binding factor 3|HDR|GATA3|GATA binding protein 3|GATA-binding factor 3
- Storage buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant mouse GATA3 (aa1 - aa258)
- Purification:Antibody is purified from culture supernatant using a protein G column
- Cat. no.:77041-580
- Packaging:Plastic vial
Rat monoclonal (KT63 ) antibody to GATA3 for WB and IF with samples derived from Mouse.
- Validated Applications: WB, IF
- Recommended Dilutions: WB: 1/500 to 1/20,000, IF: 1/100 to 1/1000
Type: Primary
Antigen: GATA3
Clonality: Monoclonal
Clone: KT63
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse