Anti-LI Cadherin Chimera Recombinant Monoclonal Antibody [clone: DMC485]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Recombinant chimeric monoclonal [DMC485] antibody to LI Cadherin for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: LI Cadherin
Clonality: Recombinant
Clone: DMC485
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 77187-458
- 77187-460
- Antigen symbol:
- LI Cadherin
- LI Cadherin
- Antigen name:
- BILL-cadherin
- BILL-cadherin
- Antigen synonyms:
- CDH16|CAD17_HUMAN|Cadherin-17|cadherin, liver-intestine|cadherin 17, LI cadherin (liver-intestine)|Cadherin 16, formerly|CDH16, formerly|Cadherin 16|Cadherin 17
- CDH16|CAD17_HUMAN|Cadherin-17|cadherin, liver-intestine|cadherin 17, LI cadherin (liver-intestine)|Cadherin 16, formerly|CDH16, formerly|Cadherin 16|Cadherin 17
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DMC485
- DMC485
- Reactivity:
- Human
- Human
- Host:
- Chimera
- Chimera
- Gene ID:
- UniprotID# Q12864
- UniprotID# Q12864
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Specifications
Related Information
Frequently Bought Together