Anti-FZD10 Chimera Recombinant Monoclonal Antibody [clone: DMC487]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Recombinant chimeric monoclonal [DMC487] antibody to FZD10 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: FZD10
Clonality: Recombinant
Clone: DMC487
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 77187-450
- 77187-452
- Antigen symbol:
- FZD10
- FZD10
- Antigen name:
- CD 350
- CD 350
- Antigen synonyms:
- FZ 10|FZD 10|Frizzled (Drosophila) homolog 10|Fz-10|CD350|CD 350 antigen|Frizzled 10|Frizzled-10|FZ10
- FZ 10|FZD 10|Frizzled (Drosophila) homolog 10|Fz-10|CD350|CD 350 antigen|Frizzled 10|Frizzled-10|FZ10
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DMC487
- DMC487
- Reactivity:
- Human
- Human
- Host:
- Chimera
- Chimera
- Gene ID:
- UniprotID# Q9ULW2
- UniprotID# Q9ULW2
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together