Anti-Calbindin Mouse Monoclonal Antibody [clone: CALB1/3333]
76967-198
- Antibody type:Primary
- Antigen name:avian-type
- Antigen symbol:Calbindin
- Clonality:Monoclonal
- Clone:CALB1/3333
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P05937
- Antigen synonyms:Calbindin|Calbindin D28|CALB|CALB1_HUMAN|CALB1|D 28K|Calbindin 1 28kDa|CALB 1|CAB27
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 7 - 96, of human CALB1 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76967-198
- Packaging:Plastic vial
Mouse monoclonal [CALB1/3333] antibody to Calbindin for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Calbindin
Clonality: Monoclonal
Clone: CALB1/3333
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human