Anti-p53 Mouse Monoclonal Antibody [clone: SPM349]
76955-054
- Antibody type:Primary
- Antigen name:Antigen NY-CO-13
- Antigen symbol:p53
- Clonality:Monoclonal
- Clone:SPM349
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Hamster,Monkey,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P04637
- Antigen synonyms:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:53 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Gel - purified p53 - beta Galactosidase fusion protein containing murine p53 from amino acids 14 - 389
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Cat. no.:76955-054
- Packaging:Plastic vial
Mouse monoclonal [SPM349] antibody to p53 for Flow Cytometry, IF, WB and IHC with samples derived from Human, Monkey, Bovine, Canine, Mouse, Rat and Hamster.
- Validated Applications: Flow Cytometry, IF, WB, IHC
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: SPM349
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey;Bovine;Dog;Mouse;Rat;Hamster