Anti-SERBP1 Mouse Monoclonal Antibody [clone: SERBP1/3492]
76961-752
- Size:100 µg
- Cat. no.:76961-752
Mouse monoclonal [SERBP1/3492] antibody to SERBP1 for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: SERBP1
Clonality: Monoclonal
Clone: SERBP1/3492
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human