Anti-14-3-3E Mouse Monoclonal Antibody [clone: CPTC-YWHAE-1]
Catalog # 76963-734
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3E
- Clonality:Monoclonal
- Clone:CPTC-YWHAE-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P62268
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:29.4 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human YWHAE protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76963-734
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CPTC-YWHAE-1] antibody to 14-3-3E for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: 14-3-3E
Clonality: Monoclonal
Clone: CPTC-YWHAE-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human