Anti-gamma Parvin Rabbit Recombinant Antibody [clone: PARVG/6312R]
76966-702
- Antibody type:Primary
- Antigen symbol:gamma Parvin
- Clonality:Recombinant
- Clone:PARVG/6312R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9HBI0
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:GST-tagged recombinant protein corresponding to human ga mMa Parvin
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76966-702
- Packaging:Plastic vial
Recombinant rabbit monoclonal [PARVG/6312R] antibody to gamma Parvin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: gamma Parvin
Clonality: Recombinant
Clone: PARVG/6312R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human