Order Entry
Canada
ContactUsLinkComponent
Anti-TDP2 Mouse Monoclonal Antibody [clone: TDP2/1258]
Anti-TDP2 Mouse Monoclonal Antibody [clone: TDP2/1258]
Catalog # 76964-082
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-TDP2 Mouse Monoclonal Antibody [clone: TDP2/1258]
Catalog # 76964-082
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A249591-100

Specifications

  • Size:
    100 µg
  • Cat. no.:
    76964-082

Specifications

About this item

Mouse monoclonal [TDP2/1258] antibody to TDP2 for Flow Cytometry, IF and IHC-P with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF, IHC-P
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: TDP2
Clonality: Monoclonal
Clone: TDP2/1258
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human