Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ESR1/3373]
Catalog # 76951-026
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:DKFZp686N23123
- Antigen symbol:Estrogen Receptor alpha
- Clonality:Monoclonal
- Clone:ESR1/3373
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P03372
- Antigen synonyms:Estradiol receptor|ER|Era|ER alpha|ESRA|ER-alpha|ESR|ESR1|ESR1_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:~67 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human Estrogen Receptor alpha protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76951-026
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ESR1/3373] antibody to Estrogen Receptor alpha for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Monoclonal
Clone: ESR1/3373
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human