Anti-Uroplakin 1A Mouse Monoclonal Antibody [clone: UPK1A/2925]
Catalog # 76964-046
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen symbol:Uroplakin 1A
- Clonality:Monoclonal
- Clone:UPK1A/2925
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00322
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:29 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 114 - 173, of human Uroplakin 1A protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76964-046
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [UPK1A/2925] antibody to Uroplakin 1A for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Uroplakin 1A
Clonality: Monoclonal
Clone: UPK1A/2925
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human