Anti-HFQ Goat Polyclonal Antibody
76833-568
- Antibody type:Primary
- Antigen symbol:HFQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Burkholderia cenocepacia
- Western blot:Yes
- Size:600 µg
- Form:Liquid
- Storage buffer:Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at 4 ℃. Avoid freeze/thaw cycles.
- Concentration:3 mg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant Burkholderia cenocepacia HFQ protein, expressed in and purified from E. coli
- Purification:Affinity purification
- Cat. no.:76833-568
- Packaging:Plastic vial
Goat polyclonal antibody to HFQ for WB with samples derived from Burkholderia cenocepacia.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: HFQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Burkholderia cenocepacia