Order Entry
Canada
ContactUsLinkComponent
Anti-SHD Rabbit Polyclonal Antibody
Anti-SHD Rabbit Polyclonal Antibody
Catalog # 77064-706
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-SHD Rabbit Polyclonal Antibody
Catalog # 77064-706
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A101591-100

Specifications

  • Size:
    100 µg
  • Cat. no.:
    77064-706

Specifications

About this item

Rabbit polyclonal antibody to SHD for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: SHD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human