Anti-GNL3L Rabbit Polyclonal Antibody
Catalog # 77080-410
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:FLJ10613
- Antigen symbol:GNL3L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NVN8
- Antigen synonyms:novel GTPase|GNL3L|Guanine nucleotide binding protein-like 3 (nucleolar)-like|OTTHUMP00000023374
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:65 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human GNL3L (amino acids 71 - 120)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77080-410
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GNL3L for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000
Type: Primary
Antigen: GNL3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human