Anti-DDX24 Rabbit Polyclonal Antibody
77072-882
- Antibody type:Primary
- Antigen name:ATP dependent RNA helicase DDX24
- Antigen symbol:DDX24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9GZR7
- Antigen synonyms:DEAD/H (Asp Glu Ala Asp) box polypeptide 24|S. cerevisiae CHL1 like helicase|DDX24_HUMAN|DDX 24|DEAD box protein 24|DEAD (Asp Glu Ala Asp) box polypeptide 24|ATP-dependent RNA helicase ddx24
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:96 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human DDX24 (amino acids 41 - 90)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77072-882
- Packaging:Plastic vial
Rabbit polyclonal antibody to DDX24 for IHC and ELISA with samples derived from Human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: DDX24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human