Anti-Vasopressin V1b receptor Rabbit Polyclonal Antibody
77079-724
- Antibody type:Primary
- Antigen name:Antidiuretic hormone receptor 1b
- Antigen symbol:Vasopressin V1b receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P47901
- Antigen synonyms:Arginine vasopressin receptor 1B|AVPR V3|AVP R3|AVPR V1b|V1bR|AVPR 1B|AVPR3|AVPR 3|Avpr1b
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:46 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human AVPR1B (amino acids 275 - 324)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77079-724
- Packaging:Plastic vial
Rabbit polyclonal antibody to AVPR1B for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: Vasopressin V1b receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human