Anti-HSPBAP1 Rabbit Polyclonal Antibody
Catalog # 77079-702
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:27 KdA heat shock protein-associated protein 1
- Antigen symbol:HSPBAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q96EW2
- Antigen synonyms:HSPBAP1|PASS1|HBAP1_HUMAN|Protein associated with small stress protein 1|HSPB1-associated protein 1|protein associating with small stress protein
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:55 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from the Internal region of human HSPBAP1 (amino acids 321 - 370)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. no.:77079-702
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to HSPBAP1 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: HSPBAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human