Anti-ZC3H11A Rabbit Polyclonal Antibody
Catalog # 77071-918
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:DKFZp686D03108
- Antigen symbol:ZC3H11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O75152
- Antigen synonyms:Zinc finger CCCH type domain containing 11A|zinc finger CCCH type containing 11A|DKFZp686F14109|Zinc finger CCCH domain containing protein 11A|ZC3HDC11A|KIAA0663|DKFZp781G2455|ZC3H11A
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:89 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ZC3H11A (amino acids 291 - 340)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77071-918
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ZC3H11A for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: ZC3H11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human