Anti-Mouse Serum Albumin Rabbit Polyclonal Antibody
77066-270
- Antibody type:Primary
- Antigen name:Albumin 1
- Antigen symbol:Mouse Serum Albumin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P02768
- Antigen synonyms:Mouse Serum Albumin|alb|Alb 1|ALBU_MOUSE|Albumin
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:69 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from the C-terminal region of human ALB (amino acids 511 - 560)
- Tested applications:IHC
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. no.:77066-270
- Packaging:Plastic vial
Rabbit polyclonal antibody to ALB for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC-P: 1:50 to 1:300, ELISA: 1:10000
Type: Primary
Antigen: Mouse Serum Albumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human