Anti-CD147 Rabbit Monoclonal Antibody [Clone: ARC0964]
Rabbit monoclonal [ARC0964] antibody to CD147 for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: CD147
Clonality: Monoclonal
Clone: ARC0964
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Frequently Bought Together