Anti-Glutathione Peroxidase 4 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Glutathione Peroxidase 4 for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:1,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Glutathione Peroxidase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together