Order Entry
Canada
ContactUsLinkComponent
 
Anti-beta Actin Rabbit Polyclonal Antibody
Catalog # 77172-560
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-beta Actin Rabbit Polyclonal Antibody
Catalog # 77172-560
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A304904-100

Specifications

  • Cat. no.:
    77172-560

Specifications

About this item

Rabbit polyclonal antibody to beta Actin for WB and ICC/IF with samples derived from Human.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100

Type: Primary
Antigen: beta Actin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human