Anti-HSP47 Mouse Monoclonal Antibody [Clone: 1C4-1A6]
Catalog # 77172-520
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:47 kDa heat shock protein
- Antigen symbol:HSP47
- Clonality:Monoclonal
- Clone:1C4-1A6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Size:200 µg
- Form:Liquid
- Gene ID:UniprotID# P50454.2
- Antigen synonyms:CBP1|Arsenic transactivated protein 3|47 kDa heat shock protein precursor|CBP 1|Arsenic-transactivated protein 3|AsTP3|AsTP 3|CBP 2|CBP2
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular weight:47 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human HSP47, full length.
- Tested applications:ICC/IF
- Purification:Protein G purification
- Cat. no.:77172-520
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [1C4-1A6] antibody to HSP47 for WB and ICC/IF with samples derived from Human.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100
Type: Primary
Antigen: HSP47
Clonality: Monoclonal
Clone: 1C4-1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human