Anti-HSP70 Rabbit Polyclonal Antibody
Catalog # 77172-408
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:DnaK type molecular chaperone HSP70 1
- Antigen symbol:HSP70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Plant,Algae
- Western blot:Yes
- Size:100 µl
- Form:Lyophilized
- Gene ID:UniprotID# A8HYV3
- Antigen synonyms:Heat shock 70 kDa protein 1|FLJ54303|FLJ75127|FLJ54408|Epididymis secretory protein Li 103|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|FLJ54370|FLJ54392
- Storage buffer:Supplied as lyophilized antiserum
- Molecular weight:~70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Mature HSP70B protein expressed with N- and C- terminal hexahistidine tags in E.coli, purified with Ni-NTA.
- Cat. no.:77172-408
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to HSP70 for WB and IP with samples derived from Algae and Plant.
- Validated Applications: WB, IP
- Recommended Dilutions: WB: 1:10,000
Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Algae, Plant