Anti-4-Hydroxynonenal Mouse Monoclonal Antibody [Clone: 12F7]
77172-196
- Antibody type:Primary
- Antigen symbol:4-Hydroxynonenal
- Clonality:Monoclonal
- Clone:12F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Species independent
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH)
- Tested applications:ICC/IF
- Purification:Protein G purification
- Cat. no.:77172-196
- Packaging:Plastic vial
Mouse monoclonal [12F7] antibody to 4-Hydroxynonenal for WB, ICC/IF and ELISA with samples derived from Species Independent.
- Validated Applications: WB, ICC/IF, ELISA
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:50, ELISA: 1:1,000
Type: Primary
Antigen: 4-Hydroxynonenal
Clonality: Monoclonal
Clone: 12F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent