Anti-STAT6 Rabbit Recombinant Antibody [Clone: RM473]
76836-516
- Cat. no.:76836-516
Rabbit monoclonal [RM473] antibody to STAT6 for IHC and WB with samples derived from Human.
- Validated Applications: IHC, WB
- Recommended Dilutions: IHC: 1:100 to 1:200, WB: 1:1,000 to 1:2,500
Type: Primary
Antigen: STAT6
Clonality: Recombinant
Clone: RM473
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human