Order Entry
Canada
ContactUsLinkComponent
Anti-Rabbit IgG H&L Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Anti-Rabbit IgG H&L Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Catalog # 76834-918
Supplier:  ANTIBODIES.COM LLC
Anti-Rabbit IgG H&L Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Catalog # 76834-918
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A294794-1

Specifications

  • Cat. no.:
    76834-918

Specifications

About this item

Goat anti-rabbit IgG H&L (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Rabbit.

  • Validated Applications: WB, IHC, ICC, ELISA
  • Recommended Dilutions: WB: 1:5,000 to 1:50,000, IHC: 1:100 to 1:1,000, ICC: 1:100 to 1:1,000, ELISA: 1:5,000 to 1:50,000

Type: Secondary
Antigen: Rabbit IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit