Order Entry
Canada
ContactUsLinkComponent
Anti-RBMS2 Mouse Monoclonal Antibody [Clone: PCRP-RBMS2-1B6]
Anti-RBMS2 Mouse Monoclonal Antibody [Clone: PCRP-RBMS2-1B6]
Catalog # 76958-518
Supplier:  Antibodies.com
Anti-RBMS2 Mouse Monoclonal Antibody [Clone: PCRP-RBMS2-1B6]
Catalog # 76958-518
Supplier:  Antibodies.com
Supplier Number:  A278359-100

Specifications

  • Cat. no.:
    76958-518

Specifications

About this item

Mouse monoclonal [PCRP-RBMS2-1B6] antibody to RBMS2 for ELISA, IP and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, IP, Flow Cytometry
  • Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells

Type: Primary
Antigen: RBMS2
Clonality: Monoclonal
Clone: PCRP-RBMS2-1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human