Anti-CD3 epsilon Rabbit Recombinant Antibody [Clone: C3e/4652R]
76962-036
- Cat. no.:76962-036
Recombinant rabbit monoclonal [C3e/4652R] antibody to CD3 epsilon for ELISA, WB and IHC-P with samples derived from Human.
- Validated Applications: ELISA, WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD3 epsilon
Clonality: Recombinant
Clone: C3e/4652R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human