Order Entry
Canada
ContactUsLinkComponent
Anti-Keratin 76 Rabbit Recombinant Antibody [Clone: KRTH/1576R]
Anti-Keratin 76 Rabbit Recombinant Antibody [Clone: KRTH/1576R]
Catalog # 76952-442
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Keratin 76 Rabbit Recombinant Antibody [Clone: KRTH/1576R]
Catalog # 76952-442
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A254190-100

Specifications

  • Cat. no.:
    76952-442

Specifications

About this item

Recombinant rabbit monoclonal [KRTH/1576R] antibody to Keratin 76 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 0.25 to 0.5 µg/ml

Type: Primary
Antigen: Keratin 76
Clonality: Recombinant
Clone: KRTH/1576R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human