Anti-BrdU Mouse Recombinant Antibody [Clone: rBRD469]
76967-192
- Cat. no.:76967-192
Recombinant mouse monoclonal [rBRD469] antibody to BrdU for IHC-P.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: BrdU
Clonality: Recombinant
Clone: rBRD469
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: