Anti-Sarcomeric Actinin alpha 2 Mouse Monoclonal Antibody [Clone: ACTN2/3292]
76949-228
- Antibody type:Primary
- Antigen symbol:Sarcomeric Actinin alpha 2
- Clonality:Monoclonal
- Clone:ACTN2/3292
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P35609
- Storage buffer:Supplied in 10mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:103 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, within amino acids 557-692, of human ACTN2 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76949-228
- Packaging:Plastic vial
Mouse monoclonal [ACTN2/3292] antibody to Sarcomeric Actinin alpha 2 for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Sarcomeric Actinin alpha 2
Clonality: Monoclonal
Clone: ACTN2/3292
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human