Anti-Endothelin A Receptor Rabbit Polyclonal Antibody
Catalog # 77052-828
Supplier: Antibodies.com
Specifications
- Antibody type:Primary
- Antigen name:Ednra
- Antigen symbol:Endothelin A Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P25101
- Antigen synonyms:Endothelin 1 receptor precursor|Endothelin-1 receptor|EDNRA_HUMAN|Endothelin receptor ET1 specific type|Endothelin ET A receptor|Endothelin receptor type A|Endothelin A Receptor|Endothelin receptor subtype A|Endothelin 1 specific receptor
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:48 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human EDNRA (amino acids 378-427)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77052-828
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to EDNRA for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IF: 1:100-1:500, ELISA: 1:5000
Type: Primary
Antigen: Endothelin A Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat