Anti-HINT1 Rabbit Polyclonal Antibody
77052-768
- Antibody type:Primary
- Antigen name:Adenosine 5' monophosphoramidase
- Antigen symbol:HINT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P49773
- Antigen synonyms:HINT1|PKCI1|PKCI 1|Adenosine 5''-monophosphoramidase|HINT 1|Histidine triad nucleotide-binding protein 1|HINT1_HUMAN|PKCI-1|Histidine triad nucleotide binding protein 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:13 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human HINT1 (amino acids 71-120)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77052-768
- Packaging:Plastic vial
Rabbit polyclonal antibody to HINT1 for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IF: 1:100-1:500, ELISA: 1:20000
Type: Primary
Antigen: HINT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat