Anti-TNFAIP8 Rabbit Polyclonal Antibody
Catalog # 77052-652
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:GG2 1
- Antigen symbol:TNFAIP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O95379
- Antigen synonyms:SCC S2|SCC-S2|TFIP8_HUMAN|NF-kappa-B-inducible DED-containing protein|MDC-3.13|TNF alpha-induced protein 8|SCCS2|Head and neck tumor and metastasis-related protein|NDED
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:23 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TFIP8 (amino acids 31-80)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77052-652
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TFIP8 for WB, IHC, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:10000
Type: Primary
Antigen: TNFAIP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse