Anti-Nicotinic Acetylcholine Receptor alpha 3 Rabbit Polyclonal Antibody
77051-416
- Antibody type:Primary
- Antigen name:ACHA3_HUMAN
- Antigen symbol:Nicotinic Acetylcholine Receptor alpha 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P32297
- Antigen synonyms:Cholinergic receptor nicotinic alpha polypeptide 3|Cholinergic receptor nicotinic alpha 3|LNCR2|Cholinergic receptor neuronal nicotinic alpha polypeptide 3|CHRNA 3|NACHRA 3|MGC104879|AChR|CHRNA3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human AChRalpha3 (amino acids 90-139)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77051-416
- Packaging:Plastic vial
Rabbit polyclonal antibody to AChRalpha3 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: Nicotinic Acetylcholine Receptor alpha 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat