Anti-CELSR2 Rabbit Polyclonal Antibody
77051-084
- Antibody type:Primary
- Antigen name:Cadherin EGF LAG seven pass G type receptor 2 flamingo Drosophila homolog
- Antigen symbol:CELSR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9HCU4
- Antigen synonyms:CELR2_HUMAN|epidermal growth factor like 2|EGFL2|EGF like domain multiple 2|Cadherin family member 10|Cadherin EGF LAG seven-pass G-type receptor 2|CDHF10|EGF-like protein 2|CELSR2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:317 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human CELSR2 (amino acids 2781-2830)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77051-084
- Packaging:Plastic vial
Rabbit polyclonal antibody to CELSR2 for IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:5000
Type: Primary
Antigen: CELSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat