Anti-GDNF Receptor alpha 1 Rabbit Polyclonal Antibody
77050-288
- Antibody type:Primary
- Antigen name:GDNF family receptor alpha 1
- Antigen symbol:GDNF Receptor alpha 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P56159
- Antigen synonyms:GDNFR|GDNF RA|GDNF R|GDNFR alpha 1|GDNF Receptor alpha 1|GDNFR alpha|GDNF receptor alpha|GDNF receptor alpha-1|GDNF family receptor alpha-1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:51 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GFR alpha-1 (amino acids 51-100)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77050-288
- Packaging:Plastic vial
Rabbit polyclonal antibody to GFR alpha-1 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:1000
Type: Primary
Antigen: GDNF Receptor alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat