Anti-NOTCH4 Rabbit Polyclonal Antibody
Catalog # 77050-132
Supplier: Antibodies.com
Specifications
- Antibody type:Primary
- Antigen name:FLJ16302
- Antigen symbol:NOTCH4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q99466
- Antigen synonyms:INT3|Neurogenic locus notch homolog protein 4|Notch homolog 4|NOTC4_HUMAN|NOTCH3|Notch 4 intracellular domain|hNotch4|hNotch 4|NOTCH4 protein
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NOTCH4 (amino acids 1401-1450)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. no.:77050-132
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NOTCH4 (cleaved Val1432) for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: NOTCH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human