Anti-MAPKAPK2/3 Rabbit Polyclonal Antibody
77049-904
- Antibody type:Primary
- Antigen name:3PK
- Antigen symbol:MAPKAPK2/3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P49137
- Antigen synonyms:MK2|AA960234|MAPK activated protein kinase 2|MAPKAP kinase 2|MAP kinase activated protein kinase 3|MAP kinase activated protein kinase 2|MAPKAP kinase 3|MAPKAPK2/3|MAPK activated protein kinase 3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human MAPKAPK2 (amino acids 169-218)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77049-904
- Packaging:Plastic vial
Rabbit polyclonal antibody to MAPKAPK2 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: MAPKAPK2/3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat