Anti-SLC4A8 + SLC4A10 Rabbit Polyclonal Antibody
77048-320
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6U841, Q2Y0W8
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:125 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC4A8/10 (amino acids 411-460)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77048-320
- Packaging:Plastic vial
Rabbit polyclonal antibody to SLC4A8 + SLC4A10 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:40000
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat