Anti-ITIH1 Rabbit Polyclonal Antibody
Catalog # 77047-800
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:H1P
- Antigen symbol:ITIH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P19827
- Antigen synonyms:Inter-alpha-trypsin inhibitor heavy chain H1|IGHEP1|Inter-alpha-trypsin inhibitor complex component III|Inter alpha trypsin inhibitor heavy chain 1|IATIH|Inter alpha (globulin) inhibitor H1|Inter-alpha-inhibitor heavy chain 1|Inter alpha trypsin inhibitor heavy chain H1|Inter alpha (globulin) inhibitor H1 polypeptide
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:71 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ITIH1 (amino acids 623-672)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77047-800
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ITIH1 (cleaved Asp672) for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: ITIH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse