Anti-COP1 Rabbit Polyclonal Antibody
Catalog # 77047-780
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Constitutive photomorphogenesis protein 1 homolog
- Antigen symbol:COP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NHY2
- Antigen synonyms:ring finger and WD repeat domain 2|E3 ubiquitin-protein ligase RFWD2|COP1|COP|RING finger and WD repeat domain protein 2|RFWD2_HUMAN|RING finger protein 200|hCOP1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:80 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RFWD2 around the phosphorylation site of Ser387 (amino acids 353-402)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. no.:77047-780
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to RFWD2 (phospho Ser387) for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:1000
Type: Primary
Antigen: COP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse