Anti-CYP2R1 Rabbit Polyclonal Antibody
Catalog # 77046-592
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:CP2R1_HUMAN
- Antigen symbol:CYP2R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6VVX0
- Antigen synonyms:CYP2R1|Cytochrome P450 family 2 R1|Vitamin D 25 hydroxylase|Cytochrome P450 2R1|Cytochrome P450 family 2 subfamily R polypeptide 1|Vitamin D 25-hydroxylase|EC 1.14.13.15|MGC4663
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Cytochrome P450 2R1 (amino acids 230-279)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77046-592
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Cytochrome P450 2R1 for WB, IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: CYP2R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse