Anti-Phospholipase C gamma 1 + 2 Rabbit Polyclonal Antibody
77045-244
- Antibody type:Primary
- Antigen name:1 phosphatidylinositol 4,5 bisphosphate phosphodiesterase gamma 1
- Antigen symbol:Phospholipase C gamma 1 + 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P19174
- Antigen synonyms:1-phosphatidyl-D-myo-inositol-4,5-bisphosphate|fj57a05|Cded|FCAS3|EC 3.1.4.11|fj46f04|APLAID|AI894140|1 phosphatidylinositol 4,5 bisphosphate phosphodiesterase gamma 2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:148 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PLCG1 around the phosphorylation site of Tyr783 (amino acids 751-800)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. no.:77045-244
- Packaging:Plastic vial
Rabbit polyclonal antibody to PLCG1 (phospho Tyr783) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:5000
Type: Primary
Antigen: Phospholipase C gamma 1 + 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat