Anti-PPAR gamma 1+2 Rabbit Polyclonal Antibody
Catalogue # 77045-116
Fournisseur: ANTIBODIES.COM LLC
Spécifications
- Antibody type:Primary
- Antigen name:NR1C3
- Antigen symbol:PPAR gamma 1+2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P37231
- Antigen synonyms:PPARG_HUMAN|PPARG|PPARG1|PPARG2|Peroxisome proliferator-activated receptor gamma|PPAR gamma|PPAR gamma 1+2|Nuclear receptor subfamily 1 group C member 3|PPAR-gamma
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PPAR-gamma around the phosphorylation site of Ser112 (amino acids 78-127)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. no.:77045-116
- Packaging:Plastic vial
Spécifications
À propos de cet article
Rabbit polyclonal antibody to PPAR-gamma (phospho Ser112) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: IF: 1:100-1:500, ELISA: 1:1000
Type: Primary
Antigen: PPAR gamma 1+2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat